Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: 293T, HeLa, BT-474, NIH/3T3, Mouse brain, Mouse kidney, Mouse liver, Rat brain. The target is usually found in the following locations: Cytoplasm, Endoplasmic reticulum membrane, Golgi apparatus membrane, Peripheral membrane protein, cytosol, perinuclear region.
Immunogen sequence: LEENGYRSLC EQVSHHPPAA AHHAESKNGW TLRQEIKITS KFRGKYLSIM PLGTIHCIFH ATGHHYTWKK VTTTVHNIIV GKLWIDQSGE IDIVNHKTGD KCNLKFVPYS YFSRDVARKV TGEVTDPSGK VHFALLGTWD EKMECFKVQP VIGENGGDAR QRGHEAEESR VMLWKRNPLP KNAENMYYFS ELALTLNAWE SGTAPTDSRL RPDQRLMENG RWDEANAEKQ RLEEKQRLSR KKREAEAMKA TEDGTPYDPY KALWFERKKD PVTKELTHIY RGEYWECKEK QDWSSCPDIF
Oxysterol binding protein is an intracellular protein that is believed to transport sterols from lysosomes to the nucleus where the sterol down-regulates the genes for the LDL receptor, HMG-CoA reductase, and HMG synthetase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:8503; Oxysterol-binding protein 1
Gene Aliases: 1110018F06Rik; AW559088; Kiaa4220; mKIAA4220; OSBP; OSBP1
UniProt ID: (Human) P22059, (Mouse) Q3B7Z2
Entrez Gene ID: (Human) 5007, (Mouse) 76303, (Rat) 365410
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support