Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MALFAVFQT TFFLTLLSLR TYQSEVLAER LPLTPVSLKV STNSTRQSLH LQWTVHNLPY HQELKMVFQI QISRIETSNV IWVGNYSTTV KWNQVLHWSW ESELPLECAT HFVRIKSLVD DAKFPEPNFW SNWSSWEEVS VQDSTGQDIL FVFPKDKLVE EGTNVTICYV SRNIQNNVSC YLEGKQIHGE QLDPHVTAFN LNSVPFIRNK GTNIYCEASQ GNVSEGMKGI VLFVSKVLEE PKDFSCETED FKTLHCTWDP GTDTALGWSK QPSQSYTLFE SFSGEKKLCT HKNWCNWQIT QDSQETYNFT LIAENYLRKR SVNILFNLTH RGETRVVTAH RGH (1-342 aa encoded by BC010943)
Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF.Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IL-31 receptor subunit beta; IL-31R subunit beta; Interleukin-31 receptor subunit beta; MGC150626; MGC150627; MGC75127; oncostatin-M specific receptor beta subunit; Oncostatin-M-specific receptor subunit beta
Gene Aliases: IL-31R-beta; IL-31RB; OSMR; OSMRB; PLCA1
UniProt ID: (Human) Q99650
Entrez Gene ID: (Human) 9180
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support