Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Description: This monoclonal AN-V2 antibody reacts with the human oncostatin M (OSM) receptor. OSM, which belongs to the interleukin-6 cytokine family, is produced by T cells, activated monocytes, and dendritic cells. In humans, OSM can bind two unique receptor complexes. The type I OSM receptor is composed of the LIFR associated with gp130, and recognizes both LIF and OSM. The type II OSM receptor, which consists of gp130 and the OSMR subunit, specifically binds the cytokine, leading to activation of the JAK/STAT signaling pathway. Reports have also indicated a soluble form of the OSM receptor, which this antibody also recognizes. The OSM receptor is expressed in the bone marrow, endothelium, liver, lung, and numerous tumor cell lines.
The AN-V2 monoclonal antibody recognizes the type II (beta) OSM receptor.
Applications Reported: This AN-V2 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This AN-V2 antibody has been pre-titrated and tested by flow cytometric analysis of the A375 cell line. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF.Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: IL-31 receptor subunit beta; IL-31R subunit beta; Interleukin-31 receptor subunit beta; MGC150626; MGC150627; MGC75127; oncostatin-M specific receptor beta subunit; Oncostatin-M-specific receptor subunit beta
Gene Aliases: IL-31R-beta; IL-31RB; OSMR; OSMRB; PLCA1
UniProt ID: (Human) Q99650
Entrez Gene ID: (Human) 9180
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support