Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: ESLHRGPPGA EPHSTHPKEP CQDLTFTNLD ALHTLSACPP NPGLVSPSPS DQREAVAIVS TPRSQWRRQC L
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Otopetrin-2; Proton channel OTOP2
Gene Aliases: OTOP2
UniProt ID: (Human) Q7RTS6
Entrez Gene ID: (Human) 92736
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support