Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.
Immunogen sequence: VKCRLGVGGS DPGGHPGDLA PNSDPVEGAT LLPGEDITTV GSTPASLAVS AKDPDKQPGP QGGPNPSQAG QQQGQQKQKR HRTRFTPAQL NELE
This gene encodes a member of the homeodomain (HD) family. HD family proteins are helix-turn-helix transcription factors that play key roles in the specification of cell fates. This protein may function during brain development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein orthopedia; orthopedia homolog
Gene Aliases: OTP
UniProt ID: (Human) Q5XKR4
Entrez Gene ID: (Human) 23440, (Rat) 294640
Molecular Function: helix-turn-helix transcription factor homeodomain transcription factor DNA-binding transcription factor gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support