Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Otubains are deubiquitylating cysteine proteases that belong to the ovarian tumor protein superfamily. Like other DUBs, otubains cleave proteins precisely at the ubiquitin -protein bond.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme OTUB2; FLJ21916; MGC3102; OTU domain, ubiquitin aldehyde binding 2; OTU domain-containing ubiquitin aldehyde-binding protein 2; Otubain-2; Ubiquitin thioesterase OTUB2; ubiquitin-specific protease otubain 2; Ubiquitin-specific-processing protease OTUB2
Gene Aliases: C14orf137; OTB2; OTU2; OTUB2
UniProt ID: (Human) Q96DC9
Entrez Gene ID: (Human) 78990
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support