Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
OTUD4 alternatively spliced transcript variants have been found for this gene. The smaller protein isoform encoded by the shorter transcript variant is found only in HIV-1 infected cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HIV-1 induced protein HIN-1; HIV-1-induced protein HIN-1; OTU domain-containing protein 4; otud4
Gene Aliases: 4930431L18Rik; AI449692; D8Ertd69e; DUBA6; HIN-1; HIN1; HSHIN1; KIAA1046; mKIAA1046; OTUD4
UniProt ID: (Human) Q01804, (Mouse) B2RRE7
Entrez Gene ID: (Human) 54726, (Mouse) 73945
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support