Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is Daudi cell lysate.
PA5-20707 can be used with blocking peptide PEP-0822.
OTUD4 alternatively spliced transcript variants have been found for this gene. The smaller protein isoform encoded by the shorter transcript variant is found only in HIV-1 infected cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HIV-1 induced protein HIN-1; HIV-1-induced protein HIN-1; OTU domain containing 4; OTU domain-containing protein 4; otud4
Gene Aliases: 4930431L18Rik; AI449692; D8Ertd69e; DUBA6; HIN-1; HIN1; HSHIN1; KIAA1046; mKIAA1046; OTUD4; RGD1305606
UniProt ID: (Human) Q01804, (Mouse) B2RRE7
Entrez Gene ID: (Human) 54726, (Mouse) 73945, (Rat) 307774
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support