Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTLDMDAVL SDFVRSTGAE PGLARDLLEG KNWDVNAALS DFEQLRQVHA GNLPPSFSEG SGGSRTPEKG FSDREPTRPP RPILQRQDDI VQEKRLSRGI SHASSSIVSL ARSHVSSNGG GGGSNEHPLE MPICAFQLPD LTVYNEDFRS FIERDLIEQS MLVALEQAGR LNWWVSVDPT SQRLLPLATT GDGNCLLHAA SLGMWGFHDR DLMLRKALYA LMEKGVEKEA LKRRWRWQQT QQNEESGLVY TEDEWQKEWN ELIKLASSEP RMHLGTNGAN CGGVESSEEP VYESLEEFHV FVLAHVLRRP IVVVADTMLR DSGGEAFAPI PFGGIYLPLE VPASQCHRSP LVLAYD (1-355 aa encoded by B C020622)
Negative regulator of the non-canonical NF-kappa-B pathway that acts by mediating deubiquitination of TRAF3, an inhibitor of the NF-kappa-B pathway, thereby acting as a negative regulator of B-cell responses. In response to non-canonical NF-kappa-B stimuli, deubiquitinates 'Lys-48'-linked polyubiquitin chains of TRAF3, preventing TRAF3 proteolysis and over-activation of non-canonical NF-kappa-B. Negatively regulates mucosal immunity against infections. Deubiquitinates ZAP70, and thereby regulates T cell receptor (TCR) signaling that leads to the activation of NF-kappa-B (PubMed:26903241). Plays a role in T cell homeostasis and is required for normal T cell responses, including production of IFNG and IL2. Mediates deubiquitination of EGFR (PubMed:22179831). Has deubiquitinating activity toward 'Lys-11', 'Lys-48' and 'Lys-63'-linked polyubiquitin chains (PubMed:27732584). Has a much higher catalytic rate with 'Lys-11'-linked polyubiquitin chains (in vitro); however the physiological significance of these data are unsure (PubMed:27732584). Hydrolyzes both linear and branched forms of polyubiquitin. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cellular zinc finger anti-NF-kappa-B protein; cellular zinc finger anti-NF-kappaB Cezanne; Cezanne; OTU domain containing 7B; OTU domain-containing protein 7B; Zinc finger A20 domain-containing protein 1; Zinc finger protein Cezanne; zinc finger, A20 domain containing 1
Gene Aliases: CEZANNE; OTUD7B; ZA20D1
UniProt ID: (Human) Q6GQQ9
Entrez Gene ID: (Human) 56957
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support