Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: YGMNGLGLAG PAMDLLHPSV GYPATPRKQR RERTTFTRSQ LDVLEALFAK TRYPDIFMRE EVALKINLPE SRVQVWFKNR RAKCRQQQQS GSGTKSRPAK KKSSPVR
Probably plays a role in the development of the brain and the sense organs. Can bind to the BCD target sequence (BTS): 5'-TCTAATCCC-3'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein OTX1; Orthodenticle homolog 1; OTX
Gene Aliases: OTX1
UniProt ID: (Human) P32242
Entrez Gene ID: (Human) 5013
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support