Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Despite its predicted molecular weight, OVGP1 often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human placenta lysate.
PA5-20580 can be used with blocking peptide PEP-0700.
Oviductins belong to a family of glycoproteins that have been suggested to play several roles during the early processes of reproduction. OVGP1 is a large carbohydrate-rich, epithelial glycoprotein with numerous O-glycosylation sites within threonine, serine, and proline-rich tandem repeats. The protein is secreted from non-ciliated oviductal epithelial cells and associates with ovulated oocytes, blastomeres, and spermatozoan acrosomal regions. At least two isoforms of OVGP are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chitinase 5; Estrogen-dependent oviduct protein; mucin 9; Mucin-9; oviduct glycoprotein; oviduct glycoprotein OGP; Oviduct-specific glycoprotein; Oviductal glycoprotein; oviductal glycoprotein 1, 120kD; oviductal glycoprotein 1, 120kDa; Oviductin
Gene Aliases: 120kDa; AU016433; AU019448; CHIT5; EGP; MOGP; MUC9; OGP; OVGP1
UniProt ID: (Human) Q15841, (Mouse) Q62010
Entrez Gene ID: (Human) 5016, (Mouse) 12659
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support