Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with bovine, canine and porcine based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene encodes a G-protein coupled receptor that binds orexin A and orexin B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hypocretin (orexin) receptor 1; hypocretin (orexin) receptor 2; hypocretin receptor 1; hypocretin receptor 2; Hypocretin receptor type 1; Hypocretin receptor type 2; hypocretin receptor-1; orexin receptor 1; orexin receptor 2; Orexin receptor type 1; Orexin receptor type 2; orexin receptor-1; orexin receptor-2; orexin type-2 receptor; Orexin/Hypocretin receptor type 1; Ox-1-R; Ox-2-R; ox1-R; ox1r; ox2-R; ox2r
Gene Aliases: HCRTR1; HCRTR2; Hctr1; mOX2aR; mOX2bR; Mox2r; mOXR2; ox-1-R; ox1-R; OX1R; OX2R
UniProt ID: (Human) O43613, (Human) O43614, (Mouse) P58307, (Rat) P56718, (Rat) P56719, (Mouse) P58308
Entrez Gene ID: (Human) 3061, (Human) 3062, (Mouse) 230777, (Rat) 25593, (Rat) 25605, (Mouse) 387285
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support