Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes a member of the 3-oxoacid CoA-transferase gene family. The encoded protein is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate. Mutations in this gene are associated with succinyl CoA:3-oxoacid CoA transferase deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-oxoacid CoA transferase 1; 3-oxoacid CoA-transferase 1; 3-oxoacid-CoA transferase 1; oxct1 {ECO:0000312|EMBL:AAI66478.1}; SCOT; SCOT-s; somatic-type succinyl CoA:3-oxoacid CoA-transferase; Somatic-type succinyl-CoA:3-oxoacid CoA-transferase; somatic-type succinyl-CoA:3-oxoacid-CoA-transferase; Succinate-coenzyme A transferase; succinyl CoA:3-oxoacid CoA transferase; succinyl-CoA:3-ketoacid CoA transferase; Succinyl-CoA:3-ketoacid coenzyme A transferase 1, mitochondrial; succinyl-CoA:3-ketoacid-CoA transferase; succinyl-CoA:3-ketoacid-coenzyme A transferase 1, mitochondrial; Succinyl-CoA:3-oxoacid CoA transferase; succinyl-CoA:alpha-ketoacid coenzyme A transferase
Gene Aliases: 2610008O03Rik; OXCT; OXCT1; Oxct2a; SCOT; Scot-s
UniProt ID: (Human) P55809, (Pig) Q29551, (Mouse) Q9D0K2, (Rat) B2GV06
Entrez Gene ID: (Human) 5019, (Rabbit) 100356813, (Pig) 396978, (Mouse) 67041, (Rat) 690163
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support