Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSEDSSALPW SINRDDYELQ EVIGSGATAV VQAAYCAPKK EKVAIKRINL EKCQTSMDEL LKEIQAMSQC HHPNIVSYYT SFVVKDELWL VMKLLSGGSV LDIIKHIVAK GEHKSGVLDE STIATILREV LEGLEYLHKN GQIHRDVKAG NILLGEDGSV QIADFGVSAF LATGGDITRN KVRKTFVGTP CWMAPEVMEQ VRGYDFKADI WSFGITAIEL ATGAAPYHKY PPMKVLMLTL QNDPPSLETG VQDKEMLKKY GKSFRKMISL CLQKDPEKRP TAAELLRHKF FQKAKNKEFL QEKTLQRAPT ISERAKKVRR VPGSSGRLHK TEDGGWEWSD DEFDEESEEG KAAISQLRSP RVKESISNSE LFPTTDPVGT LLQVPEQISA HLPQPAGQIA TQPTQVSLPP TAEPAKTAQA LSSGSGSQET KIPISLVLRL RNSKKELNDI RFEFTPGRDT AEGVSQELIS AGLVDGRDLV IVAANLQKIV EEPQSNRSVT FKLASGVEGS DIPDDGKLIG FAQLSIS
OXSR1 serine/threonine kinase or (OSR1) is activated by osmotic stresses and regulates downstream protein kinases such as PAK1 in order to establish the sensor/signaling modules that initiate the cellular response to environmental stress. OXSR1 can be phosphorylated by ATM or ATR during the DNA damage process. It regulates downstream kinases in response to environmental stress, and may play a role in regulating the actin cytoskeleton.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C6orf182; CEP57R; OTTHUMP00000209062; Oxidative stress-responsive 1 protein; oxidative-stress responsive 1; OXSR1; Serine/threonine-protein kinase OSR1
Gene Aliases: 2210022N24Rik; 2810422B09Rik; AI462649; AW209236; KIAA1101; mKIAA1101; OSR1; OXSR1
UniProt ID: (Human) O95747, (Mouse) Q6P9R2
Entrez Gene ID: (Human) 9943, (Mouse) 108737, (Rat) 316064
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support