Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Dog, Bovine, Elephant, Horse, Rabbit (100%) Gibbon, Mouse (94%) Bat (81%).
Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. This opsin gene is expressed in the eye, brain, testes, and spinal cord. This gene belongs to the seven-exon subfamily of mammalian opsin genes that includes peropsin (RRH) and retinal G protein coupled receptor (RGR). Like these other seven-exon opsin genes, this gene may encode a protein with photoisomerase activity. Alternative splicing results in multiple transcript variants encoding different isoforms. Detected in the eye, testis, brain, retina and cell lines derived from neural retina.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DJ402H5.1; G-protein coupled receptor 136; G-protein coupled receptor PGR12; Neuropsin; Opsin-5; Transmembrane protein 13
Gene Aliases: GPR136; GRP136; OPN5; PGR12; TMEM13
UniProt ID: (Human) Q6U736
Entrez Gene ID: (Dog) 100856777, (Rabbit) 100353027, (Horse) 100055886, (Bovine) 541280, (Human) 221391
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support