Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Reconstitution: 1 X 50 mcg double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
ORAI1 is a plasma membrane protein essential for store-operated calcium entry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium release-activated calcium channel protein 1; calcium release-activated calcium modulator 1; FLJ14466; Protein orai-1; Transmembrane protein 142A
Gene Aliases: CRACM1; IMD9; ORAI1; ORAT1; TAM2; TMEM142A
UniProt ID: (Human) Q96D31
Entrez Gene ID: (Human) 84876
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support