Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Control: mouse brain, rat testis
Predicted Reactivity: Human (100%), Rhesus Monkey (100%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Orexin B is a neuropeptides that play a significant role in the regulation of food intake and sleep-wakefulness, possibly by coordinating the complex behavioral and physiologic responses of these complementary homeostatic functions. A broader role in the homeostatic regulation of energy metabolism, autonomic function, hormonal balance and the regulation of body fluids, is also suggested. Orexin-A binds to both OX1R and OX2R with a high affinity, whereas orexin-B binds only to OX2R with a similar high affinity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hcrt; Hypocretin; Hypocretin (orexin) neuropeptide; Hypocretin neuropeptide precursor; orexin; orexin A; orexin B; Orexin precursor; Prepro-orexin; Preprohypocretin
Gene Aliases: HCRT; NRCLP1; orexin-A; OX; PPORX; PPOX
UniProt ID: (Human) O43612, (Mouse) O55241, (Rat) O55232
Entrez Gene ID: (Human) 3060, (Mouse) 15171, (Rat) 25723
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support