Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: KRVFGMFRQ ASDREAVYAC FTFSHWLVYA NSAANPIIYN FLSGKFREQF KAAFSCCLPG LGPCGSLKAP SPRSSASHKS LSLQSRCSIS KISEHVVLTS VTTVLP (321-425 aa encoded by BC074796)
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hcrt receptor 1; hypocretin (orexin) receptor 1; hypocretin receptor 1; Hypocretin receptor type 1; hypocretin receptor-1; Orexin a receptor; orexin receptor 1; Orexin receptor type 1; orexin receptor-1; Orexin-1 receptor; Orexin/Hypocretin receptor type 1; Ox-1-R; OX-R1; OX1 receptor; ox1-R; ox1r
Gene Aliases: HCRTR1; Hctr1; ox-1-R; ox1-R; OX1R
UniProt ID: (Human) O43613, (Mouse) P58307, (Rat) P56718
Entrez Gene ID: (Human) 3061, (Mouse) 230777, (Rat) 25593
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support