Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Positive Control - IHC: mouse tibia tissue.
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BGP; bone gamma carboxyglutamate protein 1; Bone Gla protein; Gamma-carboxyglutamic acid-containing protein; OG1; Osteocalcin; PMF1; RP11-54H19.5
Gene Aliases: Bglap; Bglap1; mOC-A; OC; Ocn; OG1
UniProt ID: (Mouse) P86546
Entrez Gene ID: (Mouse) 12096
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support