Product References
Coxsackievirus B3 infects and disrupts human induced-pluripotent stem cell derived brain-like endothelial cells.
Frontiers in cellular and infection microbiology
Mamana J,Humber GM,Espinal ER,Seo S,Vollmuth N,Sin J,Kim BJ
MA5-13854 was used in Immunocytochemistry-immunofluorescence to elucidate the unique effects of CVB3 infection on the BBB and shed light on potential mechanisms by which the virus can initiate infections in the brain.
Fri May 05 00:00:00 UTC 2023
Chemical biology fluorescent tools for in vitro investigation of the multidrug resistant P-glycoprotein (P-gp) expression in tumor cells.
RSC advances
Daumar P,Goisnard A,Dubois C,Roux M,Depresle M,Penault-Llorca F,Bamdad M,Mounetou E
MA5-13854 was used in Immunocytochemistry-immunofluorescence to report the development of 25 novel fluorescent small-molecule conjugates with varying physicochemical and optical properties, and their biological evaluation in a cell model as P-gp targeted constructs.
Fri Sep 08 00:00:00 UTC 2023
Characterization of resistant MCF-7 breast cancer cells developed by repeated cycles of photodynamic therapy.
Frontiers in pharmacology
Aniogo EC,George BP,Abrahamse H
Published figure using P-Glycoprotein monoclonal antibody (Product # MA5-13854) in Immunocytochemistry
Tue Oct 04 00:00:00 UTC 2022
Characterization of resistant MCF-7 breast cancer cells developed by repeated cycles of photodynamic therapy.
Frontiers in pharmacology
Aniogo EC,George BP,Abrahamse H
Published figure using P-Glycoprotein monoclonal antibody (Product # MA5-13854) in Immunocytochemistry
Tue Oct 04 00:00:00 UTC 2022
The Ubiquitin E3 Ligase Nedd4 Regulates the Expression and Amyloid-β Peptide Export Activity of P-Glycoprotein.
International journal of molecular sciences
Chai AB,Callaghan R,Gelissen IC
Published figure using P-Glycoprotein monoclonal antibody (Product # MA5-13854) in Western Blot
Tue Jan 18 00:00:00 UTC 2022
A microengineered Brain-Chip to model neuroinflammation in humans.
iScience
Pediaditakis I,Kodella KR,Manatakis DV,Le CY,Barthakur S,Sorets A,Gravanis A,Ewart L,Rubin LL,Manolakos ES,Hinojosa CD,Karalis K
MA5-13854 was used in Immunocytochemistry to present a human organotypic microphysiological system (MPS) that includes endothelial-like cells, pericytes, glia, and cortical neurons and maintains brain and blood-brain barrier permeability at in vivo relevant levels.
Fri Aug 19 00:00:00 UTC 2022
Mechanisms of P-Glycoprotein Regulation Under Exogenous and Endogenous Oxidative Stress In Vitro.
Acta naturae
Abalenikhina YV,Shchulkin AV,Mylnikov PY,Rokunov ED,Yakusheva EN
MA5-13854 was used in Western Blotting to investigate the mechanisms of P-glycoprotein (P-gp) transporter regulation in Caco-2 cells under exogenous and endogenous oxidative stress (OS).
Thu Nov 10 00:00:00 UTC 2022
Subcellular distribution of ezrin/radixin/moesin and their roles in the cell surface localization and transport function of P-glycoprotein in human colon adenocarcinoma LS180 cells.
PloS one
Kobori T,Tameishi M,Tanaka C,Urashima Y,Obata T
MA5-13854 was used in Flow cytometry/Cell sorting to determine the gene expression patterns and subcellular localization of ERM and P-gp and investigate the role of ERM proteins in the plasma membrane localization and transport function of P-gp using the human colon adenocarcinoma cell line LS180.
Tue Oct 26 00:00:00 UTC 2021
Subcellular distribution of ezrin/radixin/moesin and their roles in the cell surface localization and transport function of P-glycoprotein in human colon adenocarcinoma LS180 cells.
PloS one
Kobori T,Tameishi M,Tanaka C,Urashima Y,Obata T
MA5-13854 was used in Flow cytometry/Cell sorting to determine the gene expression patterns and subcellular localization of ERM and P-gp and investigate the role of ERM proteins in the plasma membrane localization and transport function of P-gp using the human colon adenocarcinoma cell line LS180.
Tue Oct 26 00:00:00 UTC 2021
Subcellular distribution of ezrin/radixin/moesin and their roles in the cell surface localization and transport function of P-glycoprotein in human colon adenocarcinoma LS180 cells.
PloS one
Kobori T,Tameishi M,Tanaka C,Urashima Y,Obata T
MA5-13854 was used in Flow cytometry/Cell sorting to determine the gene expression patterns and subcellular localization of ERM and P-gp and investigate the role of ERM proteins in the plasma membrane localization and transport function of P-gp using the human colon adenocarcinoma cell line LS180.
Tue Oct 26 00:00:00 UTC 2021
LightSpot®-FL-1 Fluorescent Probe: An Innovative Tool for Cancer Drug Resistance Analysis by Direct Detection and Quantification of the P-glycoprotein (P-gp) on Monolayer Culture and Spheroid Triple Negative Breast Cancer Models.
Cancers
Goisnard A,Daumar P,Dubois C,Aubel C,Roux M,Depresle M,Gauthier J,Vidalinc B,Penault-Llorca F,Mounetou E,Bamdad M
Published figure using P-Glycoprotein monoclonal antibody (Product # MA5-13854) in Immunocytochemistry
Wed Aug 11 00:00:00 UTC 2021
LightSpot®-FL-1 Fluorescent Probe: An Innovative Tool for Cancer Drug Resistance Analysis by Direct Detection and Quantification of the P-glycoprotein (P-gp) on Monolayer Culture and Spheroid Triple Negative Breast Cancer Models.
Cancers
Goisnard A,Daumar P,Dubois C,Aubel C,Roux M,Depresle M,Gauthier J,Vidalinc B,Penault-Llorca F,Mounetou E,Bamdad M
Published figure using P-Glycoprotein monoclonal antibody (Product # MA5-13854) in Immunocytochemistry
Wed Aug 11 00:00:00 UTC 2021
Detection and analysis of multiple biomarkers in ovarian cancer: clinical significance in diagnosis, treatment, and prognosis evaluation.
Gland surgery
Ji R,Li Y,He C,Zhu X,He A,Lu Y
MA5-13854 was used in Immunohistochemistry (Paraffin) to explore the clinical significance of CA125, CK7, CK20, ER, PR, C-erbb2, and P-gp in ovarian cancer.
Tue Dec 01 00:00:00 UTC 2020
Duodenum Intestine-Chip for preclinical drug assessment in a human relevant model.
eLife
Kasendra M,Luc R,Yin J,Manatakis DV,Kulkarni G,Lucchesi C,Sliz J,Apostolou A,Sunuwar L,Obrigewitch J,Jang KJ,Hamilton GA,Donowitz M,Karalis K
Published figure using P-Glycoprotein monoclonal antibody (Product # MA5-13854) in Immunofluorescence
Tue Jan 14 00:00:00 UTC 2020
Long noncoding RNA lncARSR confers resistance to Adriamycin and promotes osteosarcoma progression.
Cell death & disease
Shen P,Cheng Y
MA5-13854 was used in Western Blotting to illustrate the role of long noncoding RNA ARSR (lncARSR) in the development of adapted ADM resistance.
Wed May 13 00:00:00 UTC 2020
In situ molecular characterization of endoneurial microvessels that form the blood-nerve barrier in normal human adult peripheral nerves.
Journal of the peripheral nervous system : JPNS
Ouyang X,Dong C,Ubogu EE
MA5-13854 was used in Immunocytochemistry-immunoflourescence to investigate proteins specifically expressed by restrictive endoneurial microvascular endothelium in four adult sural nerves.
Sat Jun 01 00:00:00 UTC 2019
Combined effects of octreotide and cisplatin on the proliferation of side population cells from anaplastic thyroid cancer cell lines.
Oncology letters
Li Z,Jiang X,Chen P,Wu X,Duan A,Qin Y
Published figure using P-Glycoprotein monoclonal antibody (Product # MA5-13854) in Western Blot
Sat Sep 01 00:00:00 UTC 2018
The Human Blood-Nerve Barrier Transcriptome.
Scientific reports
Palladino SP,Helton ES,Jain P,Dong C,Crowley MR,Crossman DK,Ubogu EE
MA5-13854 was used in Immunocytochemistry-immunoflourescence to sequence the human blood-nerve barrier transcriptome using confluent P3 and P8 pHEndEC monolayers and normal adult laser-capture microdissected endoneurial microvessels.
Tue Dec 12 00:00:00 UTC 2017
Analysis of Cancer-Targeting Alkylphosphocholine Analogue Permeability Characteristics Using a Human Induced Pluripotent Stem Cell Blood-Brain Barrier Model.
Molecular pharmaceutics
Clark PA,Al-Ahmad AJ,Qian T,Zhang RR,Wilson HK,Weichert JP,Palecek SP,Kuo JS,Shusta EV
MA5-13854 was used in immunocytochemistry to examine trafficking of cancer-targeting alkylphosphocholine analogues across the blood brain barrier
Tue Sep 06 00:00:00 UTC 2016
Differentiation and characterization of human pluripotent stem cell-derived brain microvascular endothelial cells.
Methods (San Diego, Calif.)
Stebbins MJ,Wilson HK,Canfield SG,Qian T,Palecek SP,Shusta EV
MA5-13854 was used in immunocytochemistry to describe methods to differentiate and characterize hPSC-derived brain microvascular endothelial cells
Sun May 15 00:00:00 UTC 2016
Influence of P-glycoprotein on brucine transport at the in vitro blood-brain barrier.
European journal of pharmacology
Xu DH,Yan M,Li HD,Fang PF,Liu YW
MA5-13854 was used in immunocytochemistry to study the ability of P-glycoprotein to prevent the transport of brucine through an in vitro blood-brain barrier
Wed Sep 05 00:00:00 UTC 2012
ABCB1 is predominantly expressed in human fetal neural stem/progenitor cells at an early development stage.
Journal of neuroscience research
Yamamoto A,Shofuda T,Islam MO,Nakamura Y,Yamasaki M,Okano H,Kanemura Y
MA5-13854 was used in immunocytochemistry and immunohistochemistry to study the expression of the ABCB1 transporter in human fetal neural stem/progenitor cells
Tue Sep 01 00:00:00 UTC 2009
[The effect of resistance-related proteins on the prognosis and survival of patients with osteosarcoma: an immunohistochemical analysis].
Acta orthopaedica et traumatologica turcica
Ozger H,Eralp L,Atalar AC,Toker B,Esberk Ateş L,Sungur M,Bilgiç B,Ayan I
MA5-13854 was used in immunohistochemistry to examine the prognostic significance of multiple genes in osteosarcoma
Tue May 26 00:00:00 UTC 2009
Efficacy of Hsp90 inhibition for induction of apoptosis and inhibition of growth in cervical carcinoma cells in vitro and in vivo.
Cancer chemotherapy and pharmacology
Schwock J,Pham NA,Cao MP,Hedley DW
MA5-13854 was used in western blot to study the effect of Hsp90 inhibitors on cervical carcinoma survival and growth in vitro and in vivo
Tue Apr 01 00:00:00 UTC 2008
Effect of the novel antipsychotic drug perospirone on P-glycoprotein function and expression in Caco-2 cells.
European journal of clinical pharmacology
Zhou YG,Li KY,Li HD
MA5-13854 was used in immunocytochemistry to study the effects of the novel anti-psychotic drug perospirane of the expression and pump activity of P-glycoprotein
Tue Jul 01 00:00:00 UTC 2008
ABCG2 overexpression in colon cancer cells resistant to SN38 and in irinotecan-treated metastases.
International journal of cancer
Candeil L,Gourdier I,Peyron D,Vezzio N,Copois V,Bibeau F,Orsetti B,Scheffer GL,Ychou M,Khan QA,Pommier Y,Pau B,Martineau P,Del Rio M
MA5-13854 was used in western blot to study the role of ABCG2 overexpression in the development of irinotecan resistance in colon cancer cells
Mon May 10 00:00:00 UTC 2004
Reversal of P-glycoprotein-mediated multidrug resistance with small interference RNA (siRNA) in leukemia cells.
Cancer gene therapy
Peng Z,Xiao Z,Wang Y,Liu P,Cai Y,Lu S,Feng W,Han ZC
MA5-13854 was used in western blot to study the ability of siRNA targeted against P-glycoprotein to reverse multi-drug resistance in a human leukemia cell line
Mon Nov 01 00:00:00 UTC 2004
Involvement of extracellular signal-regulated kinase/mitogen-activated protein kinase pathway in multidrug resistance induced by HBx in hepatoma cell line.
World journal of gastroenterology
Guan J,Chen XP,Zhu H,Luo SF,Cao B,Ding L
MA5-13854 was used in western blot to investigate the role of HBx protein on the occurrence of multidrug resistance in hepatoma cell line
Wed Dec 01 00:00:00 UTC 2004
P-glycoprotein expression and distribution in the rat placenta during pregnancy.
Reproductive toxicology (Elmsford, N.Y.)
Novotna M,Libra A,Kopecky M,Pavek P,Fendrich Z,Semecky V,Staud F
MA5-13854 was used in western blot to study the expression and distribution of P-glycoprotein in the rat placenta during pregnancy
Wed Jan 26 00:00:00 UTC 2005
Differences in multidrug resistance phenotype and matrix metalloproteinases activity between endothelial cells from normal brain and glioma.
Journal of neurochemistry
Régina A,Demeule M,Bérubé A,Moumdjian R,Berthelet F,Béliveau R
MA5-13854 was used in western blot to study differences in the expression of multidrug resistance pumps and MMP activity in endothelial cells from normal brain and glioma
Wed Jan 01 00:00:00 UTC 2003
Examination of the functional activity of P-glycoprotein in the rat placental barrier using rhodamine 123.
The Journal of pharmacology and experimental therapeutics
Pavek P,Staud F,Fendrich Z,Sklenarova H,Libra A,Novotna M,Kopecky M,Nobilis M,Semecky V
MA5-13854 was used in western blot to study the functional activity of P-glycoprotein in the rat placental barrier
Sun Jun 01 00:00:00 UTC 2003
Dexamethasone modulation of multidrug transporters in normal tissues.
FEBS letters
Demeule M,Jodoin J,Beaulieu E,Brossard M,Béliveau R
MA5-13854 was used in western blot to study the effect of dexamethasone on multidrug drug transporter expression in normal rat tissues
Fri Jan 15 00:00:00 UTC 1999