Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: EGLVDYYSHI YDGPWDSYPD VLAMQFNWDG YYKEVGSAFI GSSPEFEFAL YSLCFIARPG KVCQLSLGGY PLAVRTYTWD KSTYGNGKKY IATAYIVSST
This gene encodes a protein with protease activity and is expressed in the placenta. The protein may be useful as a tumor marker. Multiple alternatively spliced transcript variants have been found for this protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 22 serine protease; 26 serine protease; endonuclease, polyU-specific; Placental protein 11; PP11; Protein endoU; Uridylate-specific endoribonuclease
Gene Aliases: ENDOU; P11; PP11; PRSS26
UniProt ID: (Human) P21128
Entrez Gene ID: (Human) 8909
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support