Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: DGACSKDSDC HAGEAVTAGN GVKTGRCLRR ENLARGTCEI FAWCPLETSS RPEEPFLKEA EDFTIFIKNH IRFPKFNFSK SNVMDVKDRS FLKSCHFGP
Mmgt2 mediates Mg(2+) transport and may provide regulated pathways for Mg(2+) transport in the Golgi and post-Golgi organelles of epithelium-derived cells. Magnesium is an essential metal, but few selective transporters have been identified. The two separate gene products, Mmgt1 and Mmgt2, comprise the family that we have termed membrane Mg(2+) transporters (MMgTs), and the proteins reside in the membrane and mediate Mg(2+) transport. The transport of Mg(2+) is saturable and studies have identified that Mmgt2 fusion proteins reside in the Golgi complex and post-Golgi vesicles, including the early endosomes in COS-7 cells transfected with the respective tagged constructs. Among related pathways for Mmgt2 are transport and binding events, in particular, transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP receptor; ATP receptor subunit; ionotropic ATP receptor P2X5; lymphoid-restricted histocompatibility antigen-1; MGC47755; P2X purinoceptor 5; P2X5; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 5; purinergic receptor P2X, ligand-gated ion channel, 5
Gene Aliases: LRH-1; P2RX5; P2X5; P2X5FL; P2X5R
UniProt ID: (Human) Q93086
Entrez Gene ID: (Human) 5026
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support