Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ARVPYTHSQ TNNKTDCRLQ NQLFIAKETT LFLAATNICM DPLIYIFLCK KFTEKLPCMQ GRKTTASSQE NHSSQTDNIT LG (253-333 aa encoded by BC041116)
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 86; G-protein coupled receptor 86; G-protein coupled receptor 94; P2Y purinoceptor 13; P2Y13; purinergic receptor P2Y G-protein coupled 13; purinergic receptor P2Y, G-protein coupled, 13
Gene Aliases: FKSG77; GPCR1; GPR86; GPR94; P2RY13; P2Y13; SP174
UniProt ID: (Human) Q9BPV8
Entrez Gene ID: (Human) 53829
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support