Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 10
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel with relatively high calcium permeability. Binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle, being responsible, for example, for sympathetic vasoconstriction in small arteries, arterioles and vas deferens. Mouse studies suggest that this receptor is essential for normal male reproductive function. It is possible that the development of selective antagonists for this receptor may provide an effective non-hormonal male contraceptive pill.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP receptor; P2X purinoceptor 1; P2X receptor, subunit 1; P2X1; P2X1 receptor; programmed cell death 3; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 1; purinergic receptor P2X, ligand-gated ion channel, 1; purinergic receptor P2X1; RP-2 protein
Gene Aliases: AI323649; BB122383; P2RX1; P2x; P2X1; P2XMR; Pdcd3; RP-2
UniProt ID: (Human) P51575, (Rat) P47824, (Mouse) Q5SRU4
Entrez Gene ID: (Human) 5023, (Rat) 25505, (Mouse) 18436
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support