Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RALNQYQAQ LGEPRPGLGP REDIQRFILR SLGEKRQLYY AMEHLGTSFK DPDPWTPAAL IPEALREKLR EDQEKRPWDH EPVKPKPLTY WKDVLLLEGV TLTQDSRQLN GSERAVLDGL LTPAECGVLL QLAKDAAGAG ARSGYRGRRS PHTPHERFEG LTVLKAAQLA RAGTVGSQGA KLLLEVSERV RTLTQAYFSP ERPLHLSFTH LVCRSAIEGE QEQRMDLSHP VHADNCVLDP DTGECWREPP AYTYRDYSGL LYLNDDFQGG DLFFTEPNAL TVTARVRPRC GRLVAFSSGV ENPHGVWAVT RGRRCALALW HTWAPEHREQ EWIEAKELLQ ESQEEEEEEE EEMPSKDPSP EPPSRRHQRV QDKTGRAPRV REEL (223-605 aa encoded by B C017217)
The protein encoded by this gene belongs to the leprecan family of proteoglycans, which function as collagen prolyl hydroxylases that are required for proper collagen biosynthesis, folding and assembly. This protein, like other family members, is thought to reside in the endoplasmic reticulum. Epigenetic inactivation of this gene is associated with breast and other cancers, suggesting that it may function as a tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gene rich cluster, B; leprecan-like 2; Leprecan-like protein 2; procollagen-proline 3-dioxygenase; Prolyl 3-hydroxylase 3; Protein B
Gene Aliases: BC016431; GRCB; HSU47926; LEPREL2; P3H3
UniProt ID: (Human) Q8IVL6, (Mouse) Q8CG70
Entrez Gene ID: (Human) 10536, (Rat) 297595, (Mouse) 14789
Molecular Function:
extracellular matrix glycoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support