Product References
Natural exosome-like nanoparticles derived from ancient medicinal insect Periplaneta americana L. as a novel diabetic wound healing accelerator.
Journal of nanobiotechnology
Liao Q,Su L,Pang L,Li J,Li H,Li J,Liu Y,Zhang J
Fri May 26 00:00:00 UTC 2023
A Novel Phenazine Analog, CPUL1, Suppresses Autophagic Flux and Proliferation in Hepatocellular Carcinoma: Insight from Integrated Transcriptomic and Metabolomic Analysis.
Cancers
Chen J,Feng D,Lu Y,Zhang Y,Jiang H,Yuan M,Xu Y,Zou J,Zhu Y,Zhang J,Ge C,Wang Y
Sun Mar 05 00:00:00 UTC 2023
A Novel Phenazine Analog, CPUL1, Suppresses Autophagic Flux and Proliferation in Hepatocellular Carcinoma: Insight from Integrated Transcriptomic and Metabolomic Analysis.
Cancers
Chen J,Feng D,Lu Y,Zhang Y,Jiang H,Yuan M,Xu Y,Zou J,Zhu Y,Zhang J,Ge C,Wang Y
Sun Mar 05 00:00:00 UTC 2023
MST4 as a novel therapeutic target for autophagy and radiosensitivity in gastric cancer.
IUBMB life
Shi X,Zou J,Wang Y,Zhao J,Ye B,Qi Q,Liu F,Hu J,Li S,Tian Y
Wed Feb 01 00:00:00 UTC 2023
MST4 as a novel therapeutic target for autophagy and radiosensitivity in gastric cancer.
IUBMB life
Shi X,Zou J,Wang Y,Zhao J,Ye B,Qi Q,Liu F,Hu J,Li S,Tian Y
Wed Feb 01 00:00:00 UTC 2023
Mesenchymal stem cell-derived exosomes ameliorate TGF-β1-induced endometrial fibrosis by altering their miRNA profile.
American journal of translational research
Zhou L,Dong L,Li H,Liu H,Yang J,Huang Z
Tue Jun 13 00:00:00 UTC 2023
MicroRNA-183-5p regulates TAR DNA-binding protein 43 neurotoxicity via SQSTM1/p62 in amyotrophic lateral sclerosis.
Journal of neurochemistry
Kim HC,Zhang Y,King PH,Lu L
Wed Mar 01 00:00:00 UTC 2023
Melatonin protects against developmental PBDE-47 neurotoxicity by targeting the AMPK/mitophagy axis.
Journal of pineal research
Dong L,Sun Q,Qiu H,Yang K,Xiao B,Xia T,Wang A,Gao H,Zhang S
Tue Aug 01 00:00:00 UTC 2023
Transcutaneous Electrical Nerve Stimulation (TENS) Alleviates Brain Ischemic Injury by Regulating Neuronal Oxidative Stress, Pyroptosis, and Mitophagy.
Mediators of inflammation
Tan Z,Dong F,Wu L,Feng Y,Zhang M,Zhang F
Fri Apr 28 00:00:00 UTC 2023
Hyperlipidemia impacts osteogenesis via lipophagy.
Bone
Ji C,Zhang Z,Xu X,Song D,Zhang D
Wed Feb 01 00:00:00 UTC 2023
Hyperlipidemia impacts osteogenesis via lipophagy.
Bone
Ji C,Zhang Z,Xu X,Song D,Zhang D
Wed Feb 01 00:00:00 UTC 2023
hASCs-derived exosomal miR-155-5p targeting TGFβR2 promotes autophagy and reduces pyroptosis to alleviate intervertebral disc degeneration.
Journal of orthopaedic translation
Chen D,Jiang X,Zou H
Wed Mar 01 00:00:00 UTC 2023
A regulatory circuit comprising the CBP and SIRT7 regulates FAM134B-mediated ER-phagy.
The Journal of cell biology
Wang X,Jiang X,Li B,Zheng J,Guo J,Gao L,Du M,Weng X,Li L,Chen S,Zhang J,Fang L,Liu T,Wang L,Liu W,Neculai D,Sun Q
Mon May 01 00:00:00 UTC 2023
Low Pi stress enhances the sensitivity of hepatocellular carcinoma to sorafenib.
Biochemical pharmacology
Bi QC,Deng ZQ,Lv YF,Liu Y,Xie CS,He YQ,Tang Q
Sat Jul 01 00:00:00 UTC 2023
Curcumin protects against doxorubicin induced oxidative stress by regulating the Keap1-Nrf2-ARE and autophagy signaling pathways.
Psychopharmacology
Liao D,Shangguan D,Wu Y,Chen Y,Liu N,Tang J,Yao D,Shi Y
Mon May 01 00:00:00 UTC 2023
Antisense lncRNA-RP11-498C9.13 promotes bladder cancer progression by enhancing reactive oxygen species-induced mitophagy.
The journal of gene medicine
Song W,Li Z,Yang K,Gao Z,Zhou Q,Li P
Fri Sep 01 00:00:00 UTC 2023
Downregulation of FAPP2 gene induces cell autophagy and inhibits PI3K/AKT/mTOR pathway in T-cell acute lymphoblastic leukemia.
Hematological oncology
Yuan T,Wang J,Shi C,Wang Y,Xia B,Xu W,Yang H,Yang Y,Ye MT,Khalid S,Liang Y,Tian C,You MJ,Wang Y
Fri Apr 01 00:00:00 UTC 2022
MED16 Promotes Tumour Progression and Tamoxifen Sensitivity by Modulating Autophagy through the mTOR Signalling Pathway in ER-Positive Breast Cancer.
Life (Basel, Switzerland)
Li H,Li K,Shu D,Shen M,Tan Z,Zhang W,Pu D,Tan W,Tang Z,Jin A,Liu S
Tue Sep 20 00:00:00 UTC 2022
Polyphyllin II induced apoptosis of NSCLC cells by inhibiting autophagy through the mTOR pathway.
Pharmaceutical biology
Jiao Y,Xin M,Xu J,Xiang X,Li X,Jiang J,Jia X
Thu Dec 01 00:00:00 UTC 2022
IL-27 regulates autophagy in rheumatoid arthritis fibroblast-like synoviocytes via STAT3 signaling.
Immunobiology
Hou J,Gao W
Fri Jul 01 00:00:00 UTC 2022
Gaseous signal molecule SO2 regulates autophagy through PI3K/AKT pathway inhibits cardiomyocyte apoptosis and improves myocardial fibrosis in rats with type II diabetes.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology
Zhao J,Wu Q,Yang T,Nie L,Liu S,Zhou J,Chen J,Jiang Z,Xiao T,Yang J,Chu C
Tue Nov 01 00:00:00 UTC 2022
SS31 Ameliorates Oxidative Stress via the Restoration of Autophagic Flux to Protect Aged Mice From Hind Limb Ischemia.
Frontiers in cardiovascular medicine
Yang Q,Li C,Chen Q
Tue May 03 00:00:00 UTC 2022
Transcription factor EB enhances autophagy and ameliorates palmitate-induced insulin resistance at least partly via upregulating AMPK activity in skeletal muscle cells.
Clinical and experimental pharmacology & physiology
Wang P,Li CG,Zhou X,Ding S
Tue Feb 01 00:00:00 UTC 2022
The alleviative effect of flavonol-type Nrf2 activator rhamnazin from Physalis alkekengi L. var. franchetii (Mast.) Makino on pulmonary disorders.
Phytotherapy research : PTR
Wu XY,Wang T,Hu HX,Zhang K,Zhao Y,Zhao BB,Lou HX,Wang XN,Shen T
Fri Apr 01 00:00:00 UTC 2022
(±)-5-bromo-2-(5-fluoro-1-hydroxyamyl) Benzoate Protects Against Oxidative Stress Injury in PC12 Cells Exposed to H2O2 Through Activation of Nrf2 Pathway.
Frontiers in pharmacology
Qi S,Zhang X,Fu Z,Pi A,Shi F,Fan Y,Zhang J,Xiao T,Shang D,Lin M,Gao N,Chang J,Gao Y
Tue Aug 09 00:00:00 UTC 2022
Ginsenoside Rh4 Inhibits Colorectal Cancer Cell Proliferation by Inducing Ferroptosis via Autophagy Activation.
Evidence-based complementary and alternative medicine : eCAM
Wu Y,Pi D,Chen Y,Zuo Q,Zhou S,Ouyang M
Sat Jul 16 00:00:00 UTC 2022
Ginsenoside Rh4 Inhibits Colorectal Cancer Cell Proliferation by Inducing Ferroptosis via Autophagy Activation.
Evidence-based complementary and alternative medicine : eCAM
Wu Y,Pi D,Chen Y,Zuo Q,Zhou S,Ouyang M
Sat Jul 16 00:00:00 UTC 2022
Knocking Down Peroxiredoxin 6 Aggravates Cerebral Ischemia-Reperfusion Injury by Enhancing Mitophagy.
Neuroscience
Hong T,Zhou Y,Peng L,Wu X,Li Y,Li Y,Zhao Y
Tue Feb 01 00:00:00 UTC 2022
TPI1 activates the PI3K/AKT/mTOR signaling pathway to induce breast cancer progression by stabilizing CDCA5.
Journal of translational medicine
Jin X,Wang D,Lei M,Guo Y,Cui Y,Chen F,Sun W,Chen X
Wed May 04 00:00:00 UTC 2022
TPI1 activates the PI3K/AKT/mTOR signaling pathway to induce breast cancer progression by stabilizing CDCA5.
Journal of translational medicine
Jin X,Wang D,Lei M,Guo Y,Cui Y,Chen F,Sun W,Chen X
Wed May 04 00:00:00 UTC 2022
Apigenin alleviated PA-induced pyroptosis by activating autophagy in hepatocytes.
Food & function
Meng Z,Zhu B,Gao M,Wang G,Zhou H,Lu J,Guan S
Mon May 23 00:00:00 UTC 2022
CHMP2B regulates TDP-43 phosphorylation and cytotoxicity independent of autophagy via CK1.
The Journal of cell biology
Deng X,Sun X,Yue W,Duan Y,Hu R,Zhang K,Ni J,Cui J,Wang Q,Chen Y,Li A,Fang Y
Mon Jan 03 00:00:00 UTC 2022
Long non-coding RNA IGFBP7-AS1 promotes odontogenic differentiation of stem cells from human exfoliated deciduous teeth through autophagy: An in vitro study.
Archives of oral biology
Wang D,Zhu N,Xie F,Qin M,Wang Y
Thu Sep 01 00:00:00 UTC 2022
Overexpression of VPS16 correlates with tumor progression and chemoresistance in colorectal cancer.
Biochemical and biophysical research communications
Zhang B,Ma Y,Niu H,Liu Z
Sat Jun 04 00:00:00 UTC 2022
Autophagy Inhibition Enhances the Anti-Tumor Activity of Methylseleninic Acid in Cisplatin-Resistance Human Lung Adenocarcinoma Cells.
Frontiers in pharmacology
Xin M,Gao Q,Xiang X,Xu J,Jiao Y,Li X,Zhang X,Jia X
Wed Nov 01 00:00:00 UTC 2023
Xuesaitong Combined with Dexmedetomidine Improves Cerebral Ischemia-Reperfusion Injury in Rats by Activating Keap1/Nrf2 Signaling and Mitophagy in Hippocampal Tissue.
Oxidative medicine and cellular longevity
Han GJ,Min XZ,Ma SS,Ding C,Wang XQ
Tue Dec 20 00:00:00 UTC 2022
Zinc Oxide Nanoparticle Improves the Intestinal Function of Intrauterine Growth Retardation Finishing Pigs via Regulating Intestinal Morphology, Inflammation, Antioxidant Status and Autophagy.
Frontiers in veterinary science
Zhou B,Zhang J,Liu H,Chen S,Wang T,Wang C
Sat Jul 16 00:00:00 UTC 2022
SIRT1/mTOR pathway-mediated autophagy dysregulation promotes Pb-induced hepatic lipid accumulation in HepG2 cells.
Environmental toxicology
Zhao Y,Mao A,Zhang R,Guan S,Lu J
Tue Mar 01 00:00:00 UTC 2022
Magnetic hyperthermia induces effective and genuine immunogenic tumor cell death with respect to exogenous heating.
Journal of materials chemistry. B
Yan B,Liu C,Wang S,Li H,Jiao J,Lee WSV,Zhang S,Hou Y,Hou Y,Ma X,Fan H,Lv Y,Liu X
Wed Jul 20 00:00:00 UTC 2022
Acrylamide induces intrinsic apoptosis and inhibits protective autophagy via the ROS mediated mitochondrial dysfunction pathway in U87-MG cells.
Drug and chemical toxicology
Deng L,Zhao M,Cui Y,Xia Q,Jiang L,Yin H,Zhao L
Tue Nov 01 00:00:00 UTC 2022
Inhibition of autophagy potentiates the cytotoxicity of the irreversible FGFR1-4 inhibitor FIIN-2 on lung adenocarcinoma.
Cell death & disease
Jia X,Xin M,Xu J,Xiang X,Li X,Jiao Y,Wang L,Jiang J,Pang F,Zhang X,Zhang J
Tue Aug 30 00:00:00 UTC 2022
Resveratrol alleviates enterotoxigenic Escherichia coli K88-induced damage by regulating SIRT-1 signaling in intestinal porcine epithelial cells.
Food & function
Luo X,Wu S,Jia H,Si X,Song Z,Zhai Z,Bai J,Li J,Yang Y,Wu Z
Mon Jul 04 00:00:00 UTC 2022
GNIP1 functions both as a scaffold protein and an E3 ubiquitin ligase to regulate autophagy in lung cancer.
Cell communication and signaling : CCS
Zhou F,Liu Y,Ai W,Wang Y,Gan M,Jiang Q,Han T,Wang JB
Tue Aug 30 00:00:00 UTC 2022
Oncostatin M receptor regulates osteoblast differentiation via extracellular signal-regulated kinase/autophagy signaling.
Stem cell research & therapy
Zhou J,Yang J,Dong Y,Shi Y,Zhu E,Yuan H,Li X,Wang B
Tue Jun 28 00:00:00 UTC 2022
Protocatechualdehyde Rescues Oxygen-Glucose Deprivation/Reoxygenation-Induced Endothelial Cells Injury by Inducing Autophagy and Inhibiting Apoptosis via Regulation of SIRT1.
Frontiers in pharmacology
Cao S,Chen S,Qiao X,Guo Y,Liu F,Ding Z,Jin B
Sat May 07 00:00:00 UTC 2022
Moxidectin induces autophagy arrest in colorectal cancer.
Medical oncology (Northwood, London, England)
Mao Y,Xie H,Shu D,Cheng L,Lan J,Pan K
Thu Sep 29 00:00:00 UTC 2022
Inhibition of Nrf2 degradation alleviates age-related osteoporosis induced by 1,25-Dihydroxyvitamin D deficiency.
Free radical biology & medicine
Yang R,Zhang J,Li J,Qin R,Chen J,Wang R,Goltzman D,Miao D
Sat Jan 01 00:00:00 UTC 2022
Lipopolysaccharide reduces melanin synthesis in vitiligo melanocytes by regulating autophagy.
Experimental dermatology
Sun L,Sun J,Huo X,Feng Q,Li Y,Xie X,Geng S
Sat Oct 01 00:00:00 UTC 2022
Regulation of the sensitivity of hepatocarcinoma cells by ORMDL3, to sorafenib by autophagy.
Medical oncology (Northwood, London, England)
Sun Y,Guan X,Zhang T,Li Y,Shi H,Chitakunye AT,Hong H,Zhang S,Zhu Q,Cai L
Tue Aug 16 00:00:00 UTC 2022
Exosomes-derived miR-125-5p from cartilage endplate stem cells regulates autophagy and ECM metabolism in nucleus pulposus by targeting SUV38H1.
Experimental cell research
Chen D,Jiang X
Sun May 01 00:00:00 UTC 2022
Yifei sanjie Pills Alleviate Chemotherapy-Related Fatigue by Reducing Skeletal Muscle Injury and Inhibiting Tumor Growth in Lung Cancer Mice.
Evidence-based complementary and alternative medicine : eCAM
Wu Y,Pi D,Chen Y,Zuo Q,Lin L,Ouyang M
Wed Sep 07 00:00:00 UTC 2022
Artemisinin Alleviates Cerebral Ischemia/Reperfusion-Induced Oxidative Damage via Regulating PHB2-Mediated Autophagy in the Human Neuroblastoma SH-SY5Y Cell Line.
Oxidative medicine and cellular longevity
Jiang M,Lai X,Zhang Y,Shen M,Ma H,Liu A,Wu J,Yan J
Tue Dec 27 00:00:00 UTC 2022
Extracellular vesicle miR-32 derived from macrophage promotes arterial calcification in mice with type 2 diabetes via inhibiting VSMC autophagy.
Journal of translational medicine
Cao J,Chen C,Chen Q,Gao Y,Zhao Z,Yuan Q,Li A,Yang S,He Y,Zu X,Liu J
Wed Jul 06 00:00:00 UTC 2022
Autophagy and apoptosis mediated nano-copper-induced testicular damage.
Ecotoxicology and environmental safety
Chen H,Wang Y,Luo J,Kang M,Hou J,Tang R,Zhao L,Shi F,Ye G,He X,Cui H,Guo H,Li Y,Tang H
Sat Jan 01 00:00:00 UTC 2022
Reduction of Autophagosome Overload Attenuates Neuronal Cell Death After Traumatic Brain Injury.
Neuroscience
Quan X,Song L,Zheng X,Liu S,Ding H,Li S,Xu G,Li X,Liu L
Thu Apr 15 00:00:00 UTC 2021
Reduction of Autophagosome Overload Attenuates Neuronal Cell Death After Traumatic Brain Injury.
Neuroscience
Quan X,Song L,Zheng X,Liu S,Ding H,Li S,Xu G,Li X,Liu L
Thu Apr 15 00:00:00 UTC 2021
IL-6 regulates autophagy and chemotherapy resistance by promoting BECN1 phosphorylation.
Nature communications
Hu F,Song D,Yan Y,Huang C,Shen C,Lan J,Chen Y,Liu A,Wu Q,Sun L,Xu F,Hu F,Chen L,Luo X,Feng Y,Huang S,Hu J,Wang G
Tue Jun 15 00:00:00 UTC 2021
Selective STAT3 Inhibitor Alantolactone Ameliorates Osteoarthritis via Regulating Chondrocyte Autophagy and Cartilage Homeostasis.
Frontiers in pharmacology
Pei W,Huang X,Ni B,Zhang R,Niu G,You H
Sat Oct 16 00:00:00 UTC 2021
Gastrodin ameliorates learning and memory impairment in rats with vascular dementia by promoting autophagy flux via inhibition of the Ca2+/CaMKII signal pathway.
Aging
Chen TT,Zhou X,Xu YN,Li Y,Wu XY,Xiang Q,Fu LY,Hu XX,Tao L,Shen XC
Wed Mar 10 00:00:00 UTC 2021
Gossypol, a novel modulator of VCP, induces autophagic degradation of mutant huntingtin by promoting the formation of VCP/p97-LC3-mHTT complex.
Acta pharmacologica Sinica
Li XJ,Zhang YY,Fu YH,Zhang H,Li HX,Li QF,Li HL,Tan RK,Jiang CX,Jiang W,Li ZX,Luo C,Lu BX,Dang YJ
Fri Oct 01 00:00:00 UTC 2021
RabGEF1 functions as an oncogene in U251 glioblastoma cells and is involved in regulating AKT and Erk pathways.
Experimental and molecular pathology
Fan H,Xin T,Dong X,Yang F,Zhang R,Feng S,He D,Guo H,Pang Q
Mon Feb 01 00:00:00 UTC 2021
Elevation of Hyaluronan Synthase by Magnesium Supplementation Mediated through the Activation of GSK3 and CREB in Human Keratinocyte-Derived HaCaT Cells.
International journal of molecular sciences
Marunaka K,Shu S,Kobayashi M,Goto M,Katsuta Y,Yoshino Y,Ikari A
Wed Dec 22 00:00:00 UTC 2021
SARS-CoV-2 Causes Mitochondrial Dysfunction and Mitophagy Impairment.
Frontiers in microbiology
Shang C,Liu Z,Zhu Y,Lu J,Ge C,Zhang C,Li N,Jin N,Li Y,Tian M,Li X
Tue Jan 25 00:00:00 UTC 2022
UM-6 induces autophagy and apoptosis via the Hippo-YAP signaling pathway in cervical cancer.
Cancer letters
Wang D,He J,Dong J,Wu S,Liu S,Zhu H,Xu T
Thu Oct 28 00:00:00 UTC 2021
UM-6 induces autophagy and apoptosis via the Hippo-YAP signaling pathway in cervical cancer.
Cancer letters
Wang D,He J,Dong J,Wu S,Liu S,Zhu H,Xu T
Thu Oct 28 00:00:00 UTC 2021
Protectin DX Attenuates Lumbar Radicular Pain of Non-compressive Disc Herniation by Autophagy Flux Stimulation via Adenosine Monophosphate-Activated Protein Kinase Signaling.
Frontiers in physiology
Zhao QX,Wang YH,Wang SC,Xue S,Cao ZX,Sun T
Tue Jan 25 00:00:00 UTC 2022
Protectin DX Attenuates Lumbar Radicular Pain of Non-compressive Disc Herniation by Autophagy Flux Stimulation via Adenosine Monophosphate-Activated Protein Kinase Signaling.
Frontiers in physiology
Zhao QX,Wang YH,Wang SC,Xue S,Cao ZX,Sun T
Tue Jan 25 00:00:00 UTC 2022
GPR109A alleviate mastitis and enhances the blood milk barrier by activating AMPK/Nrf2 and autophagy.
International journal of biological sciences
Guo W,Li W,Su Y,Liu S,Kan X,Ran X,Cao Y,Fu S,Liu J
Mon Dec 20 00:00:00 UTC 2021
HORMAD1 promotes docetaxel resistance in triple negative breast cancer by enhancing DNA damage tolerance.
Oncology reports
Zong B,Sun L,Peng Y,Wang Y,Yu Y,Lei J,Zhang Y,Guo S,Li K,Liu S
Thu Jul 01 00:00:00 UTC 2021
Ginsenoside Rb1 alleviates liver injury induced by 3-chloro-1,2-propanediol by stimulating autophagic flux.
Journal of food science
Lu J,Lu J,Bu X,Li Y,Ge G,Guan S
Wed Dec 01 00:00:00 UTC 2021
PPT1 Reduction Contributes to Erianin-Induced Growth Inhibition in Oral Squamous Carcinoma Cells.
Frontiers in cell and developmental biology
Luo Q,Li X,Gan G,Yang M,Chen X,Chen F
Sat Jul 16 00:00:00 UTC 2022
Oxidative stress-induced mitophagy is suppressed by the miR-106b-93-25 cluster in a protective manner.
Cell death & disease
Zhang C,Nie P,Zhou C,Hu Y,Duan S,Gu M,Jiang D,Wang Y,Deng Z,Chen J,Chen S,Wang L
Wed Feb 24 00:00:00 UTC 2021
Menthol Targeting AMPK Alleviates the Inflammatory Response of Bovine Mammary Epithelial Cells and Restores the Synthesis of Milk Fat and Milk Protein.
Frontiers in immunology
Liu S,Guo W,Jia Y,Ye B,Liu S,Fu S,Liu J,Hu G
Mon Feb 14 00:00:00 UTC 2022
Pathogenic mechanism of congenital cataract caused by the CRYBA1/A3-G91del variant and related intervention strategies.
International journal of biological macromolecules
Xu J,Wang H,Wu C,Wang A,Wu W,Xu J,Luo C,Ni S,Yao K,Chen X
Sun Oct 31 00:00:00 UTC 2021
XPA serves as an autophagy and apoptosis inducer by suppressing hepatocellular carcinoma in a PI3K/Akt/mTOR dependent manner.
Journal of gastrointestinal oncology
Deng Y,Chen QS,Huang WF,Dai JW,Wu ZJ
Sun Aug 01 00:00:00 UTC 2021
Upstream open reading frame with NOTCH2NLC GGC expansion generates polyglycine aggregates and disrupts nucleocytoplasmic transport: implications for polyglycine diseases.
Acta neuropathologica
Zhong S,Lian Y,Luo W,Luo R,Wu X,Ji J,Ji Y,Ding J,Wang X
Wed Dec 01 00:00:00 UTC 2021
Upstream open reading frame with NOTCH2NLC GGC expansion generates polyglycine aggregates and disrupts nucleocytoplasmic transport: implications for polyglycine diseases.
Acta neuropathologica
Zhong S,Lian Y,Luo W,Luo R,Wu X,Ji J,Ji Y,Ding J,Wang X
Wed Dec 01 00:00:00 UTC 2021
Inhibition of autophagy with Chloroquine enhanced apoptosis induced by 5-aminolevulinic acid-photodynamic therapy in secondary hyperparathyroidism primary cells and organoids.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zeng L,Zou Q,Huang P,Xiong L,Cheng Y,Chen Q,Li Y,He H,Yi W,Wei W
Fri Oct 01 00:00:00 UTC 2021
Autocrined leptin promotes proliferation of non-small cell lung cancer (NSCLC) via PI3K/AKT and p53 pathways.
Annals of translational medicine
Wang J,Zhou F,Li F,Wang B,Hu Y,Li X
Thu Apr 01 00:00:00 UTC 2021
Cyanidin-3-O-glucoside and its metabolite protocatechuic acid ameliorate 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) induced cytotoxicity in HepG2 cells by regulating apoptotic and Nrf2/p62 pathways.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Zhao L,Zhou N,Zhang H,Pan F,Ai X,Wang Y,Hao S,Wang C
Mon Nov 01 00:00:00 UTC 2021
Bidirectional role of reactive oxygen species during inflammasome activation in acrolein-induced human EAhy926 cells pyroptosis.
Toxicology mechanisms and methods
Jiang L,Luo S,Qiu T,Li Q,Jiang C,Sun X,Yang G,Zhang C,Liu X,Jiang L
Mon Nov 01 00:00:00 UTC 2021
Sanguinarine impairs lysosomal function and induces ROS-dependent mitophagy and apoptosis in human hepatocellular carcinoma cells.
Archives of pharmacal research
Wang J,Su Q,Wu Q,Chen K,Ullah A,Ghauri MA,Zhang Y
Mon Nov 01 00:00:00 UTC 2021
Sanguinarine impairs lysosomal function and induces ROS-dependent mitophagy and apoptosis in human hepatocellular carcinoma cells.
Archives of pharmacal research
Wang J,Su Q,Wu Q,Chen K,Ullah A,Ghauri MA,Zhang Y
Mon Nov 01 00:00:00 UTC 2021
Collusion of α-Synuclein and Aβ aggravating co-morbidities in a novel prion-type mouse model.
Molecular neurodegeneration
Lloyd GM,Dhillon JS,Gorion KM,Riffe C,Fromholt SE,Xia Y,Giasson BI,Borchelt DR
Thu Sep 09 00:00:00 UTC 2021
TAF1L promotes development of oral squamous cell carcinoma via decreasing autophagy-dependent apoptosis.
International journal of biological sciences
Wang D,Qi H,Zhang H,Zhou W,Li Y,Li A,Liu Q,Wang Y
Thu Sep 30 00:00:00 UTC 2021
The c-Myc/miR-27b-3p/ATG10 regulatory axis regulates chemoresistance in colorectal cancer.
Theranostics
Sun W,Li J,Zhou L,Han J,Liu R,Zhang H,Ning T,Gao Z,Liu B,Chen X,Ba Y
Wed Apr 14 00:00:00 UTC 2021
In vivo and in vitro studies using Clonorchis sinensis adult-derived total protein (CsTP) on cellular function and inflammatory effect in mouse and cell model.
Parasitology research
Shang M,Sun H,Wu Y,Gong Y,Tang Z,Meng F,He L,Yu X,Huang Y,Li X
Fri May 01 00:00:00 UTC 2020
In vivo and in vitro studies using Clonorchis sinensis adult-derived total protein (CsTP) on cellular function and inflammatory effect in mouse and cell model.
Parasitology research
Shang M,Sun H,Wu Y,Gong Y,Tang Z,Meng F,He L,Yu X,Huang Y,Li X
Fri May 01 00:00:00 UTC 2020
Transcriptomic Analysis of Glycolysis-Related Genes Reveals an Independent Signature of Bladder Carcinoma.
Frontiers in genetics
Mou Z,Yang C,Zhang Z,Wu S,Xu C,Cheng Z,Dai X,Chen X,Ou Y,Jiang H
Tue Apr 19 00:00:00 UTC 2022
Niacin Alleviates Dairy Cow Mastitis by Regulating the GPR109A/AMPK/NRF2 Signaling Pathway.
International journal of molecular sciences
Guo W,Liu J,Li W,Ma H,Gong Q,Kan X,Cao Y,Wang J,Fu S
Fri May 08 00:00:00 UTC 2020
Niacin Alleviates Dairy Cow Mastitis by Regulating the GPR109A/AMPK/NRF2 Signaling Pathway.
International journal of molecular sciences
Guo W,Liu J,Li W,Ma H,Gong Q,Kan X,Cao Y,Wang J,Fu S
Fri May 08 00:00:00 UTC 2020
Sirtuin6 (SIRT6) Promotes the EMT of Hepatocellular Carcinoma by Stimulating Autophagic Degradation of E-Cadherin.
Molecular cancer research : MCR
Han LL,Jia L,Wu F,Huang C
Fri Nov 01 00:00:00 UTC 2019
Phenotypic Drug Screening for Dysferlinopathy Using Patient-Derived Induced Pluripotent Stem Cells.
Stem cells translational medicine
Kokubu Y,Nagino T,Sasa K,Oikawa T,Miyake K,Kume A,Fukuda M,Fuse H,Tozawa R,Sakurai H
Tue Oct 01 00:00:00 UTC 2019
Adiponectin inhibits high glucose-induced angiogenesis via inhibiting autophagy in RF/6A cells.
Journal of cellular physiology
Li R,Du J,Yao Y,Yao G,Wang X
Fri Nov 01 00:00:00 UTC 2019
Parkin truncating variants result in a loss-of-function phenotype.
Scientific reports
Santos M,Morais S,Pereira C,Sequeiros J,Alonso I
Wed Nov 06 00:00:00 UTC 2019
hnRNPK modulates selective quality-control autophagy by downregulating the expression of HDAC6 in 293 cells.
International journal of oncology
Li Z,Liu X,Ma J,Zhang T,Gao X,Liu L
Thu Nov 01 00:00:00 UTC 2018
Rab11a is required for porcine reproductive and respiratory syndrome virus induced autophagy to promote viral replication.
Biochemical and biophysical research communications
Wang K,Li S,Worku T,Hao X,Yang L,Zhang S
Sat Oct 14 00:00:00 UTC 2017
Rab11a is required for porcine reproductive and respiratory syndrome virus induced autophagy to promote viral replication.
Biochemical and biophysical research communications
Wang K,Li S,Worku T,Hao X,Yang L,Zhang S
Sat Oct 14 00:00:00 UTC 2017
Autophagy is essential for flavopiridol‑induced cytotoxicity against MCF‑7 breast cancer cells.
Molecular medicine reports
Wang S,Wang K,Wang H,Han J,Sun H
Fri Dec 01 00:00:00 UTC 2017
Clinicopathological description of two cases with SQSTM1 gene mutation associated with frontotemporal dementia.
Neuropathology : official journal of the Japanese Society of Neuropathology
Kovacs GG,van der Zee J,Hort J,Kristoferitsch W,Leitha T,Höftberger R,Ströbel T,Van Broeckhoven C,Matej R
Mon Feb 01 00:00:00 UTC 2016
Clinicopathological description of two cases with SQSTM1 gene mutation associated with frontotemporal dementia.
Neuropathology : official journal of the Japanese Society of Neuropathology
Kovacs GG,van der Zee J,Hort J,Kristoferitsch W,Leitha T,Höftberger R,Ströbel T,Van Broeckhoven C,Matej R
Mon Feb 01 00:00:00 UTC 2016
Parkin-induced ubiquitination of Mff promotes its association with p62/SQSTM1 during mitochondrial depolarization.
Acta biochimica et biophysica Sinica
Gao J,Qin S,Jiang C
Wed Jul 01 00:00:00 UTC 2015