Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PAAF1 inhibits proteasome 26S assembly and proteolytic activity by impairing the association of the 19S regulatory complex with the 20S core. In case of HIV-1 infection, recruited by viral Tat to the HIV-1 promoter, where it promotes the recruitment of 19S regulatory complex through dissociation of the proteasome 26S. This presumably promotes provirus transcription efficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Proteasomal ATPase-associated factor 1; Protein G-16; WD repeat domain 71; WD repeat-containing protein 71
Gene Aliases: PAAF; PAAF1; Rpn14; WDR71
UniProt ID: (Human) Q9BRP4
Entrez Gene ID: (Human) 80227
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support