Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ISLDVDADR DGVVEKNNPK KASWTWGPEG QGAILLVNCD RETPWLPKED CRDEKVYSKE DLKDMSQMIL RTKGPDRLPA GYEIVLYISM SDSDKVGVFY VENPFFGQRY IHILGRRKLY HVVKYTGGSA ELLFFVEGLC FPDEGFSGLV SIHVSLLEYM AQDIPLTPIF TDTVIFRIAP WIMTPNILPP VSVFVCCMKD NYLFLKEVKN LVEKTNCELK VCFQYLNRGD RWIQDEIEFG YIEAPHKGFP VVLDSPRDGN LKDFPVKELL GPDFGYVTRE PLFESVTSLD SFGNLEVSPP VTVNGKTYPL GRILIGSSFP L (118-438 aa encoded by BC009701)
This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type II enzyme is the most widely expressed family member. Known substrates for this enzyme include myelin basic protein in the central nervous system and vimentin in skeletal muscle and macrophages. This enzyme is thought to play a role in the onset and progression of neurodegenerative human disorders, including Alzheimer disease and multiple sclerosis, and it has also been implicated in glaucoma pathogenesis. This gene exists in a cluster with four other paralogous genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAD type II; PAD-H19; peptidyl arginine deiminase, type 2; peptidyl arginine deiminase, type II; peptidyl arginine deiminase, type II; PAD type II; Peptidylarginine deiminase II; Protein-arginine deiminase type II; Protein-arginine deiminase type-2; RP1-37C10.6
Gene Aliases: KIAA0994; mKIAA0994; PAD-H19; PAD2; PADI2; Pdi; PDI2
UniProt ID: (Human) Q9Y2J8, (Rat) P20717, (Mouse) Q08642
Entrez Gene ID: (Pig) 100621356, (Human) 11240, (Rat) 29511, (Mouse) 18600
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support