Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QLINTRQPQA KIIVLGLLPR GEKPNPLRQK NAKVNQLLKV SLPKLANVQL LDTDG
Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Platelet-activating factor (PAF) is a biologically active phospholipid with diverse biologic effects. PAF is degraded to inactive products by hydrolysis of the acetyl group at the sn-2 position to produce the biologically inactive products LYSO-PAF and acetate. This reaction is catalyzed by PAF acetylhydrolase (PAFAH). The various monomeric and multimeric forms of the enzyme are composed of alpha, beta, and gamma PAFAH subunits.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 303; intracellular platelet-activating factor acetylhydrolase alpha 2 subunit; PAF acetylhydrolase 30 kDa subunit; PAF-AH 30 kDa subunit; PAF-AH subunit beta; PAF-AH1b alpha 2 subunit; PAFAH subunit beta; pl; platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa); Platelet-activating factor acetylhydrolase IB subunit alpha2; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa)
Gene Aliases: HEL-S-303; PAFAH1B2; PAFAHB
UniProt ID: (Human) Q29459
Entrez Gene ID: (Human) 5049
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support