Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: WIPFRRVEE GEKEFHVRNP QVHQRVSECL RVLKILQEVT AGQTVFNILP GGLDLMTLKG NIDMSRVAVM GHSFGGATAI LALAKETQFR CAVALDAWMF PLERDFYPKA RGPVFFINTE KFQTMESVNL MKKICAQHEQ SRIITVLGSV HRSQTDFAFV TGNLIGKFFS TETRGSLDPY EGQEVMVRAM LAFLQKHLDL KEDYNQWNNL IEGIGPSLTP GAPHHLSSL (165-392 aa encoded by BC001158)
Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAF:lysophospholipid transacetylase; PAF:sphingosine transacetylase; platelet-activating factor acetylhydrolase 2 40kDa; platelet-activating factor acetylhydrolase 2, 40kDa; Platelet-activating factor acetylhydrolase 2, cytoplasmic; Platelet-activating factor acetyltransferase PAFAH2; SD-PLA2; Serine-dependent phospholipase A2
Gene Aliases: 2310074E22Rik; AI507170; HSD-PLA2; PAFAH2
UniProt ID: (Human) Q99487, (Rat) P83006, (Mouse) E9QNW7
Entrez Gene ID: (Human) 5051, (Rat) 313611, (Mouse) 100163
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support