Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • PAG1 Antibodies

            Invitrogen

            PAG1 Recombinant Rabbit Monoclonal Antibody (24GB685)

            View all (12) PAG1 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite PAG1 Recombinant Rabbit Monoclonal Antibody (24GB685)

            • Antibody Testing Data (3)
            Application
            PAG1 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            PAG1 Antibody (MA5-57149) in ICC/IF

            Immunocytochemical staining of PAG1 in HT-1080 cells using PAG1 Recombinant Monoclonal Antibody (Product # MA5-57149) at a 1:1,1000 dilution. Nuclei were stained blue with DAPI; PAG1 was stained magenta with Alexa Fluor® 647. Images were taken using Leica stellaris 5. Protein abundance based on laser Intensity and smart gain: High. Scale bar: 20 μm.
            View Product

            PAG1 Antibody in Immunocytochemistry (ICC/IF)
            PAG1 Antibody in Western Blot (WB)
            PAG1 Antibody in Flow Cytometry (Flow)

            Product Details

            MA5-57149

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:1,000-1:5,000
            -

            Immunocytochemistry (ICC/IF)

            1:100-1:1,000
            -

            Flow Cytometry (Flow)

            1:2,000
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Expression System

            HEK293F

            Class

            Recombinant Monoclonal

            Type

            Antibody

            Clone

            24GB685

            Immunogen

            A synthesized peptide derived from human PAG1 (aa 400-432)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.4, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Ambient (domestic); Dry ice (international)

            RRID

            AB_3680174

            Target Information

            PAG (phosphoprotein associated with GEMs), also known as Cbp (Csk-binding protein), is a ubiquitously expressed 46 kDa transmembrane adaptor protein present in membrane rafts (glycosphingolipid-enriched microdomains), which however migrates on SDS PAGE gels anomalously as an 80 kDa molecule. Following tyrosine phosphorylation by Src family kinases, PAG binds and thereby activates the protein tyrosine kinase Csk, the major negative regulator of the Src family kinases. Signaling via the B-cell receptor in B cells or high affinity IgE receptor (FcepsilonRI) in mast cells leads to PAG increased tyrosine phosphorylation and Csk binding, while T cell receptor signaling causes PAG dephosphorylation, loss of Csk binding and increased activation of the protein tyrosine kinase Lck.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: Csk binding protein; Csk-binding protein; phosphoprotein associated with glycosphingolipid microdomains 1; Phosphoprotein associated with glycosphingolipid-enriched microdomains 1; phosphoprotein transmembrane adaptor 1; phosphoprotein-associated with GEMs; Transmembrane adapter protein PAG; transmembrane adaptor protein PAG; Transmembrane phosphoprotein Cbp

            View more View less

            Gene Aliases: CBP; F730007C19Rik; PAG; PAG1

            View more View less

            UniProt ID: (Human) Q9NWQ8, (Rat) Q9JM80, (Mouse) Q3U1F9

            View more View less

            Entrez Gene ID: (Human) 55824, (Rat) 64019, (Mouse) 94212

            View more View less

            Function(s)

            transmembrane receptor protein tyrosine kinase adaptor activity SH3/SH2 adaptor activity protein binding SH2 domain binding

            Process(es)

            adaptive immune response signal transduction transmembrane receptor protein tyrosine kinase signaling pathway epidermal growth factor receptor signaling pathway positive regulation of signal transduction intracellular signal transduction T cell receptor signaling pathway regulation of T cell activation negative regulation of T cell activation immune system process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0