Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
PAK7 (KIAA1264) is implicated in the regulation of cytoskeletal dynamics, proliferation, and cell survival signaling. This kinase contains a CDC42/Rac1 interactive binding motif, and has been shown to bind CDC42 in the presence of GTP. This kinase is predominantly expressed in brain. It is capable of promoting neurite outgrowth, and thus may play a role in neurite development. This kinase is associated with microtubule networks and induces microtubule stabilization. The subcellular localization of this kinase is tightly regulated during cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p21 (RAC1) activated kinase 7; p21 protein (Cdc42/Rac)-activated kinase 7; p21(CDKN1A)-activated kinase 7; p21-activated kinase 5; p21-activated kinase 7; p21CDKN1A-activated kinase 7; PAK 5; PAK 7; PAK-5; PAK-7; PAK5; protein kinase PAK5; RP5-1119D9.3; Serine/threonine-protein kinase PAK 5; serine/threonine-protein kinase PAK7
Gene Aliases: KIAA1264; PAK5; PAK7
UniProt ID: (Human) Q9P286
Entrez Gene ID: (Human) 57144
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support