Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
PAMR1 may be involved in regeneration of skeletal muscle. Although related to peptidase S1 family, lacks the conserved active Ser residue in position 665 which is replaced by a Thr, suggesting that it has no protease activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inactive serine protease PAMR1; Peptidase domain-containing protein associated with muscle regeneration 1; regeneration associated muscle protease; Regeneration-associated muscle protease homolog
Gene Aliases: 5930437L24; DKFZP586H2123; E430002G05Rik; FP938; PAMR1; RAMP; RGD1308745; UNQ699/PRO1344
UniProt ID: (Human) Q6UXH9, (Mouse) Q8BU25
Entrez Gene ID: (Human) 25891, (Mouse) 210622, (Rat) 311252
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support