Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat
This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Two alternatively spliced transcript variants for this gene have been described. However, the full length nature of one of the variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AN2 polyA specific ribonuclease subunit homolog; Inactive ubiquitin carboxyl-terminal hydrolase 52; PAB-dependent poly(A)-specific ribonuclease subunit 2; PAB-dependent poly(A)-specific ribonuclease subunit PAN2; PAB1P-dependent poly(A)-nuclease; PAB1P-dependent poly(A)-specific ribonuclease; PABP-dependent poly(A) nuclease 2; PAN deadenylation complex catalytic subunit 2; PAN deadenylation complex subunit 2; PAN2 poly(A) specific ribonuclease subunit homolog; PAN2 polyA specific ribonuclease subunit; PAN2-PAN3 deadenylation complex catalytic subunit PAN2; Poly(A)-nuclease deadenylation complex subunit 2; ubiquitin specific peptidase 52; ubiquitin specific protease 52
Gene Aliases: 1200014O24Rik; AI047843; AW742773; KIAA0710; mKIAA0710; PAN2; USP52
UniProt ID: (Human) Q504Q3, (Mouse) Q8BGF7
Entrez Gene ID: (Human) 9924, (Mouse) 103135
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support