Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAKGDSTN VDKLVKDIYG GDYERFGLQG SAVASSFGNM MSKEKRDSIS KEDLARATLV TITNNIGSIA RMCALNENID RVVFVGNFLR INMVSMKLLA YAMDFWSKGQ LKALFLEHEG YFGAVGALLE LFKMTDDK (462-598 aa encoded by BC026296)
PANK1 belongs to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPanK; hPanK1; mPank; Pantothenate kinase 1; Pantothenic acid kinase 1
Gene Aliases: PANK; PANK1; Pank1b
UniProt ID: (Human) Q8TE04, (Mouse) Q8K4K6
Entrez Gene ID: (Human) 53354, (Mouse) 75735, (Rat) 294088
Molecular Function: kinase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support