Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FDHSGKDTERE HEPPYEISVQ EEITARLHFI KFENTYIEAC LDFIKDHLVN TETKVIQATG GGAYKFKDLI EEKLRLKVDK EDVMTCLIKG CNFVLKNIPH EAFVYQKDSD PEFRFQTNHP HIFPYLLVNI GSGVSIVKVE TE
Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is most abundant in muscle but is expressed in all tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4'-phosphopantetheine phosphatase; DKFZp547M242; FLJ10782; hPanK4; Inactive pantothenic acid kinase 4; pantothenic acid kinase 4
Gene Aliases: PANK4
UniProt ID: (Human) Q9NVE7
Entrez Gene ID: (Human) 55229
Molecular Function: kinase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support