Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SNANIDRSPQ NDDHSDTDSE ENRDNQQFLT TVKLANAKQT TEDEQAREAK SHQKCSKSCD PGEDCASCQQ DEIDVVPESP LSDVGSEDVG TGPKNDNKLT RQESCLGNSP PFEKESEPES PMDVDNSKNS CQDSEADEET SPGFDEQEDG SSSQTANKPS RFQARDADIE FRKRYSTKGG EVRLHFQFEG GESRTGMNDL NAKLPGNISS LNVECRNSKQ HGKKDSKITD HFMRLPKAED RRKEQWETKH QRTERKIPK
Poly(ADP-ribose) glycohydrolase (PARG) is the major enzyme responsible for the catabolism of poly(ADP-ribose), a reversible covalent-modifier of chromosomal proteins. The protein is found in many tissues and may be subject to proteolysis generating smaller, active products.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochondrial poly(ADP-ribose) glycohydrolase; poly (ADP-ribose) glycohydrolase; Poly(ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase 60 kDa isoform
Gene Aliases: PARG; PARG99
UniProt ID: (Human) Q86W56
Entrez Gene ID: (Human) 8505
Molecular Function: glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support