Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MEIIRSNFK SNLHKVYQAI EEADFFAIDG EFSGISDGPS VSALTNGFDT PEERYQKLKK HSMDFLLFQF GLCTFKYDYT DSKYITKSFN FYVFPKPFNR SSPDVKFVCQ SSSIDFLASQ GFDFNKVFRN GIPYLNQEEE RQLREQYDEK RSQANGAGAL SYVSPNTSKC PVTIPEDQKK FIDQVVEKIE DLLQSEENKN LDLEPCTGFQ RKLIYQTLSW KYPKGIHVET LETEKKERYI VISKVDEEER KRREQQKHAK EQEELNDAVG FSRVIHAIAN SGKLVIGHNM LLDVMHTVHQ FYCPLPADLS EFKEMTTCVF PRLLDTKLMA STQPFKDIIN NTSLAELE (1-347 aa encoded by BC050029)
The protein encoded by this gene is a 3'-exoribonuclease, with similarity to the RNase D family of 3'-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deadenylating nuclease; Deadenylation nuclease; poly(A)-specific ribonuclease (deadenylation nuclease); Poly(A)-specific ribonuclease PARN; Polyadenylate-specific ribonuclease
Gene Aliases: 1200003I18Rik; DAN; DKCB6; PARN; PFBMFT4; RGD1565449
UniProt ID: (Human) O95453, (Mouse) Q8VDG3
Entrez Gene ID: (Human) 5073, (Mouse) 74108, (Rat) 360464
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support