Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a 3'-exoribonuclease, with similarity to the RNase D family of 3'-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deadenylating nuclease; Deadenylation nuclease; Poly(A)-specific ribonuclease PARN; Polyadenylate-specific ribonuclease
Gene Aliases: DAN; DKCB6; PARN; PFBMFT4
UniProt ID: (Human) O95453
Entrez Gene ID: (Human) 5073
Molecular Function: mRNA polyadenylation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support