Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QEEAAALRQ ALTLSLLEQP PLEAEEPPDG GTDGKAQLVV HSAFEQDVEE LDRALRAALE VHVQEETVGP WRRTLPAELR ARLERCHGVS VALRGDCTIL RGFGAHPARA ARHLVALLAG PWDQSLAFPL AASGPTLAGQ TLKGPWNNLE RLAENTGEFQ EVVRAFYDTL DAARSSIRVV RVERVSHPLL QQQYELYRER LLQRCERRPV EQVLYHGTTA PAVPDICAHG FNRSFCGRNA TVYGKGVYFA RRASLSVQDR YSPPNADGHK AVFVARVLTG DYGQGRRGLR APPLRGPGHV LLRYDSAVDC ICQPSIFVIF HDTQALPTHL ITCEHVPRAS PDDPSGLPGR SPDT (673-1025 aa encoded by BC136542)
Poly(ADP-ribose) polymerases (PARPs), such as PARP10, regulate gene transcription by altering chromatin organization by adding ADP-ribose to histones. PARPs can also function as transcriptional cofactors. Poly(ADP-ribose) polymerases (PARPs), such as PARP10, regulate gene transcription by altering chromatin organization by adding ADP-ribose to histones. PARPs can also function as transcriptional cofactors (Yu et al., 2005).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 10; ARTD10; FLJ14464; PARP-10; poly (ADP-ribose) polymerase family, member 10; Poly [ADP-ribose] polymerase 10; Protein mono-ADP-ribosyltransferase PARP10
Gene Aliases: ARTD10; PARP10
UniProt ID: (Human) Q53GL7
Entrez Gene ID: (Human) 84875
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support