Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MAPKPKPWVQ TEGPEKKKGR QAGREEDPFR STAEALKAIP AEKRIIRVDP TCPLSSNPGT QVYEDYNCTL NQTNIENNNN KFYIIQLLQD SNRFFTCWNR WGRVGEVGQS KINHFTRLED AKKDFEKKFR EKTKNNWAER DHFVSHPGKY TLIEVQAEDE AQEAVVKVDR GPVRTVTKRV QPCSLDPATQ KLITNIFSKE MFKNTMALMD LDVKKMPLGK LSKQQIARGF EALEALEEAL KGPTDGGQSL EELSSHFYTV IPHNFGHSQP PPINSPELLQ AKKDMLLVLA DIELAQALQA VSEQEKTVEE VPHPLDRDYQ LLKCQLQLLD SGAPEYKVIQ TYLEQTGSNH RCPTLQHIWK VNQEGEEDRF QAHSKLGNRK LLWHGTNMAV VAAILTSGLR IMPHSGGRVG KGIYFASENS KSAGYVIGMK CGAHHVGYMF LGEVALGREH HINTDNPSLK SPPPGFDSVI ARGHTEPDPT QDTELELDGQ QVVVPQGQPV PCPEFSSSTF SQSEYLIYQE SQCRLRYLLE VHL
PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintenance of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 3; ADP-ribosyltransferase diphtheria toxin-like 3; ADPRT-3; ARTD3; DNA ADP-ribosyltransferase PARP3; IRT1; NAD(+) ADP-ribosyltransferase 3; NAD+ ADP-ribosyltransferase 3; pADPRT-3; PARP-3; poly (ADP-ribose) polymerase family, member 3; Poly [ADP-ribose] polymerase 3; poly(ADP-ribose) synthetase-3; Poly[ADP-ribose] synthase 3; poly[ADP-ribose] synthetase 3; Protein mono-ADP-ribosyltransferase PARP3
Gene Aliases: ADPRT3; ADPRTL2; ADPRTL3; ARTD3; IRT1; PADPRT-3; PARP3
UniProt ID: (Human) Q9Y6F1
Entrez Gene ID: (Human) 10039
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support