Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RALTSNCENY NTVSPSQLLD FLSGKQYAVG DETDLSIPTS PTSKYNRDNE KVQLLARKII FSYLNLLVNS KNDLAVA
Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 53%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PARI; PARP-1 binding protein; PARP1-binding protein; PARPBP; PCNA-interacting partner
Gene Aliases: AROM; C12orf48; PARI; PARPBP
UniProt ID: (Human) Q9NWS1
Entrez Gene ID: (Human) 55010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support