Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, Jurkat, Raji.
Predicted reactivity: Mouse (84%), Rat (83%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a putative member of the class II family of aminoacyl-tRNA synthetases. These enzymes play a critical role in protein biosynthesis by charging tRNAs with their cognate amino acids. This protein is encoded by the nuclear genome but is likely to be imported to the mitochondrion where it is thought to catalyze the ligation of proline to tRNA molecules. Mutations have been found in this gene in some patients with Alpers syndrome. [provided by RefSeq, Mar 2015]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Probable proline--tRNA ligase, mitochondrial; proline tRNA ligase 2, mitochondrial (putative); Prolyl-tRNA synthetase; Prolyl-tRNA synthetase 2, mitochondrial; ProRS
Gene Aliases: MT-PRORS; PARS2; proRS
UniProt ID: (Human) Q9H6S5
Entrez Gene ID: (Human) 25973
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support