Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSAPRSPT PRPRRMKKDE SFLGKLGGTL ARKRRAREVS DLQEEGKNAI NSPMSPALAD VHPEDTQLEE NEERTMIDPT SKEDPKFKEL VKVLLDWIND VLVEERIIVK QLEEDLYDGQ VLQKLLEKLA GCKLNVAEVT QSEIGQKQKL QTVLEAVHDL LRPRGWALRW SVDSIHGKNL VAILHLLVSL AMHFRAPIRL PEHVTVQVVV VRKREGLLHS SHISEELTTT TEMMMGRFER DAFDTLFDHA PDKLSVVKKS LITFVNKHLN KLNLEVTELE TQFADGVYLV LLMGLLEDYF VPLHHFYLTP ESFDQKVHNV SFAFELMLDG GLKKPKARPE DVVNLDLKST LRVLYNLFTK YKNVE (1-364 aa encoded by BC046103)
Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts. Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Affixin; Beta-parvin; parvin, beta
Gene Aliases: affixin; AI595373; AW742462; CGI-56; D15Gsk1; PARVB
UniProt ID: (Human) Q9HBI1, (Mouse) Q9ES46
Entrez Gene ID: (Human) 29780, (Mouse) 170736, (Rat) 362973
Molecular Function:
actin and actin related protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support