Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LQSTDPYLDD FYYQNYFEKL EKLSAAEEIQ GDGPKKERTK LITPQVAKLE HAYKPVQFEG SLGKLTVSSV NNPRKMIDAV VTSRSEDDET KEKQVRDKRR KTLVIIEKTY SLLLDVEDYE RRYLLSLEEE RPALMDDRKH KICSMYDNLR GKLPGQERPS DDHFVQIMCI RKGKRMVARI LPFLSTEQAA DILMTTARNL PFLIKKDAQD EVLPCLLSPF SLLLYHLPSV SITSLLRQLM NLPQSAATPA LSNPHLTAVL QNKFGLSLLL ILLSRGEDLQ SSDPATESTQ NNQWTEVMFM ATRELLRIPQ AALAKPISIP TNLVSLFSRY VDRQKLNLLE TKLQLVQGIR
RNA-binding protein involved in deadenylation-dependent decapping of mRNAs, leading to the degradation of mRNAs. Acts as a scaffold protein that connects deadenylation and decapping machinery. Required for cytoplasmic mRNA processing body (P-body) assembly. In case of infection, required for translation and replication of hepatitis C virus (HCV).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAT1-like protein 1; Pat1b; protein associated with topoisomerase II homolog 1; Protein PAT1 homolog 1; Protein PAT1 homolog b
Gene Aliases: hPat1b; OK/KNS-cl.5; Pat1b; PATL1
UniProt ID: (Human) Q86TB9
Entrez Gene ID: (Human) 219988
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support