Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MPHNSIRSGH GGLNQLGGAF VNGRPLPEVV RQRIVDLAHQ GVRPCDISRQ LRVSHGCVSK ILGRYYETGS IRPGVIGGSK PKVATPKVVE KIGDYKRQNP TMFAWEIRDR LLAEGVCDND TVPSVSSINR IIRTKVQQPF NLPMDSCVAT KSLSPGHTLI PSSAVTPPES PQSDSLGSTY SINGLLGIAQ PGSDKRKMDD SDQDSCRLSI DS
This gene encodes a member of the paired box family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:8622; paired box gene 8; Paired box protein Pax-8; paired domain gene 8
Gene Aliases: Pax-8; PAX8
UniProt ID: (Human) Q06710, (Mouse) Q00288
Entrez Gene ID: (Human) 7849, (Mouse) 18510
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support