Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: RSITDQVSDS SPYHSPKVEE WSSLGRNNFP AAAPHAVNGL EKGALEQEAK YGQAPNGLPA VGSFVSASSM APYPTPAQVS PYMTYSAAPS GYVAGH
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 9 gene is unknown but it may involve development of stratified squamous epithelia as well as various organs and skeletal elements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: paired box gene 9; Paired box protein Pax-9; paired domain gene 9; pax9 {ECO:0000312|EMBL:BAE79272.1}
Gene Aliases: PAX9; STHAG3
UniProt ID: (Human) P55771, (Rat) Q2L4T2
Entrez Gene ID: (Human) 5083, (Rat) 362741
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support