Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEGISNFKT PSKLSEKKKS VLCSTPTINI PASPFMQKLG FGTGVNVYLM KRSPRGLSHS PWAVKKINPI CNDHYRSVYQ KRLMDEAKIL KSLHHPNIVG YRAFTEANDG SLCLAMEYGG EKSLNDLIEE RYKASQDPFP AAIILKVALN MARGLKYLHQ EKKLLHGDIK SSNVVIKGDF ETIKICDVGV SLPLDENMTV TDPEACYIGT EPWKPKEAVE ENGVITDKAD IFAFGLTLWE MMTLSIPHIN LSNDDDDEDK TFDESDFDDE AYYAALGTRP PINMEELDES YQKVIELFSV CTNEDPKDRP SAAHIVEALE TDV (1-322 aa encoded by BC015191)
PBK (TOPK) is a serine/threonine kinase that is related to the dual specific mitogen-activated protein kinase (MAPKK). The kinase is up-regulated in a variety of hematological malignancies, participates in activating lymphoid cells, and supports testicular function (spermatogenesis). Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 84; CT84; epididymis luminal protein 164; Lymphokine-activated killer T-cell-originated protein kinase; MAPKK-like protein kinase; Nori-3; PDZ-binding kinase; serine/threonine protein kinase; Spermatogenesis-related protein kinase; SPK; T-LAK cell-originated protein kinase
Gene Aliases: 2810434B10Rik; AW538537; CT84; D14Ertd732e; HEL164; Nori-3; PBK; SPK; TOPK
UniProt ID: (Human) Q96KB5, (Mouse) Q9JJ78
Entrez Gene ID: (Human) 55872, (Mouse) 52033, (Rat) 290326
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support