Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
PBX2 is a ubiquitously expressed member of the TALE/PBX homeobox family. PBX2 gene was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. PBX2 is a transcriptional activator which binds to the TLX1 promoter. This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeobox 12; Homeobox protein PBX2; pre-B-cell leukemia homeobox 2; Pre-B-cell leukemia transcription factor 2; Protein G17; XXbac-BPG300A18.13
Gene Aliases: G17; HOX12; PBX2; PBX2MHC
UniProt ID: (Human) P40425
Entrez Gene ID: (Human) 5089
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support