Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is HepG2 cell lysate.
PA5-20704 can be used with blocking peptide PEP-0819.
Protocadherins comprise the largest group within the cadherin family of calcium-dependent cell-cell adhesion molecules. Protocadherin 18 (PCDH18) was initially identified through screening of human and mouse ESTs using conserved cytoplasmic domain motifs. Tissue screening revealed PCDH18 broad expression in tissues such as brain, liver, heart, kidney, lung, and trachea. PCDH18 expression is also temporally and spatially regulated in the mouse embryonic brain and interacts with Disabled-1, an intracellular adapter protein involved in neuronal migration and cell positioning during mammalian brain development. PCDH18 was also found to be expressed in the developing zebrafish neural tube and central nervous system, lending support to the hypothesis that PCDH18 may play a role during brain development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP434B0923; Protocadherin 18; protocadherin 68-like protein; Protocadherin-18
Gene Aliases: 3110038E07Rik; BB095589; KIAA1562; PCDH18; PCDH68L
UniProt ID: (Human) Q9HCL0, (Mouse) Q8VHR0
Entrez Gene ID: (Human) 54510, (Mouse) 73173, (Rat) 295027
Molecular Function: cadherin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support